Inlet liners
Split, single taper liner with glass wool, deactivated, 870 μl, 4 mm
Katalog # HEWL5183-4712
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
- Consistent high level of inertness
Specifikationer
- Ø indv.:4 mm
- Beskrivelse:Split, single taper liner with glass wool, deactivated, 870 μl
- Seq:22