Inlet liners
Straight, direct inlet liner, non-deactivated (for gas samples headspace or purge & trap), 140 μl, 1,5 mm
Katalog # HEWL5183-4710
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
- Consistent high level of inertness
Specifikationer
- Ø indv.:1,5 mm
- Beskrivelse:Straight, direct inlet liner, non-deactivated (for gas samples headspace or purge & trap), 140 μl