Capillary column ferrules for gas chromatography
Graphpack 2M connector for column ID 0,33/0,25 mm
Catalog # HEWL5182-9761
Supplier: AGILENT HEWLETT PACKARD
Capillary column ferrules for gas chromatography
Catalog # HEWL5182-9761
Supplier: AGILENT HEWLETT PACKARD
Jump to:
Product Details & Documents
Specifications
- Beskrivelse:Graphpack 2M connector for column ID 0,33/0,25 mm
- Seq:39