Orders LinkContactUsLinkComponent
Split/splitless inlet seals
Seal, gold plated with cross, use with total flow rates >200 ml/min
Katalog # HEWL5182-9652
Leverandør:   AGILENT HEWLETT PACKARD
Split/splitless inlet seals
Katalog # HEWL5182-9652
Split/splitless inlet seals
Split/splitless inlet seals
Seal, gold plated with cross, use with total flow rates >200 ml/min
Price
575,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These inlet seals have been developed to ensure that you have a consistent and inert surface to properly seal the inlet and prevent sample degradation.
  • Very reproducible smooth surface, eliminating traces of machining grooves that can be the source of minute leaks
  • High quality, leak-free seal
 : An inlet washer is provided with each inlet seal.
Specifikationer
  • Beskrivelse:
    Seal, gold plated with cross, use with total flow rates >200 ml/min
  • Seq:
    3