Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Deactivated/silinised vials

Deactivated/silinised vials

Vials, 1,00 ml, 12×32 mm, No, Crimp/snap
Catalog # HEWL5182-0567
Deactivated/silinised vials
Catalog # HEWL5182-0567
Deactivated/silinised vials
Deactivated/silinised vials

Vials, 1,00 ml, 12×32 mm, No, Crimp/snap

Price
217,00 kr
Log in for availability
1 * 100 Items
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents

These vials made of PP are used for pH sensitive samples, sodium or heavy metals analysis.

  • Translucent
  • Good chemical resistance
  • Can be used with crimp or snap caps

Vials with glass insert are used for economical and effective microsampling. The PP body has a glass flanged insert molded to the inside, so the sample comes in contact only with the 100 µl glass insert and the septum.

Specifications
  • Kasket inkluderet:
    No
  • Diameter×H:
    12×32 mm
  • Kapacitet:
    1,00 ml
  • Beskrivelse:
    Vials
  • Låg type:
    Crimp/snap
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.