Certified microvolume inserts for wide opening vials
Inserts, with polymer feet, graduated to 300 µl in increments of 100 µl, 0,25 ml, Conical bottom, PP, No
Katalog # HEWL5182-0549
Leverandør: AGILENT HEWLETT PACKARD
Certified microvolume inserts for wide opening vials
Katalog # HEWL5182-0549
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Specifikationer
- Kasket inkluderet:No
- Kapacitet:0,25 ml
- Beskrivelse:Inserts, with polymer feet, graduated to 300 µl in increments of 100 µl
- Materiale:PP
- Seq:08
- Grundlag:Conical bottom