Inlet liners
Splitless, straight liner, deactivated, quartz, 250 µl, 2 mm
Katalog # HEWL5181-8818
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
- Consistent high level of inertness
Specifikationer
- Ø indv.:2 mm
- Beskrivelse:Splitless, straight liner, deactivated, quartz, 250 µl
- Seq:40
- Cat. No.:HEWL5181-8818