Capillary column ferrules for gas chromatography
High performance ferrules, 100% vespel, short, ferrule ID 0,4 mm, column ID 0,1, 0,2, 0,25 mm
HEWL5181-3322
- Beskrivelse:High performance ferrules, 100% vespel, short, ferrule ID 0,4 mm, column ID 0,1, 0,2, 0,25 mm
- Seq:15