Orders LinkContactUsLinkComponent
Capillary column ferrules for gas chromatography
Specialty ferrules VG-1 for MS, 85% vespel, 15% graphite, without holes
Katalog # HEWL5181-3308
Leverandør:   AGILENT HEWLETT PACKARD
Capillary column ferrules for gas chromatography
Katalog # HEWL5181-3308
Capillary column ferrules for gas chromatography
Capillary column ferrules for gas chromatography
Specialty ferrules VG-1 for MS, 85% vespel, 15% graphite, without holes
Price
597,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Specialty ferrules VG-1 for MS, 85% vespel, 15% graphite, without holes
  • Seq:
    18