Capillary column ferrules for gas chromatography
Specialty ferrules VG-1 for MS, 85% vespel, 15% graphite, without holes
Katalog # HEWL5181-3308
Leverandør: AGILENT HEWLETT PACKARD
Capillary column ferrules for gas chromatography
Katalog # HEWL5181-3308
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Specialty ferrules VG-1 for MS, 85% vespel, 15% graphite, without holes
- Seq:18