Certified microvolume inserts for wide opening vials
Inserts, with polymer feet, 0,25 ml, Conical bottom, Glas, No
Katalog # 548-0748
Leverandør: AGILENT HEWLETT PACKARD
Certified microvolume inserts for wide opening vials
Katalog # 548-0748
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Specifikationer
- Kasket inkluderet:No
- Kapacitet:0,25 ml
- Beskrivelse:Inserts, with polymer feet
- Materiale:Glas
- Seq:01
- Grundlag:Conical bottom