Orders LinkContactUsLinkComponent
Capillary column ferrules for gas chromatography
Capillary column ferrules for gas chromatography
General purpose graphite ferrules, short, ferrule ID 0,5 mm, column ID 0,1, 0,2, 0,25, 0,32 mm
Catalog # HEWL5080-8853
Capillary column ferrules for gas chromatography
Catalog # HEWL5080-8853
Capillary column ferrules for gas chromatography
Capillary column ferrules for gas chromatography
General purpose graphite ferrules, short, ferrule ID 0,5 mm, column ID 0,1, 0,2, 0,25, 0,32 mm
Price
553,00 kr
Log in for availability
1 * 10 Items
Product Details & Documents
Documents
Specifications
  • Beskrivelse:
    General purpose graphite ferrules, short, ferrule ID 0,5 mm, column ID 0,1, 0,2, 0,25, 0,32 mm
  • Seq:
    04