Orders LinkContactUsLinkComponent
Capillary column ferrules for gas chromatography
General purpose graphite ferrules, short, ferrule ID 1 mm, column ID 0,53 mm
  HEWL5080-8773
Capillary column ferrules for gas chromatography
  HEWL5080-8773
Capillary column ferrules for gas chromatography
Capillary column ferrules for gas chromatography
General purpose graphite ferrules, short, ferrule ID 1 mm, column ID 0,53 mm
kr448.00
1 * 10 Items
  • Beskrivelse:
    General purpose graphite ferrules, short, ferrule ID 1 mm, column ID 0,53 mm
  • Seq:
    03