Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Forensic anion solutions kit

Forensic anion solutions kit

Buffer, basic anion, for CE, pH 12,1, 50 ml, indirect UV-detection buffer, 1
Katalog # HEWL5064-8209
Leverandør:   AGILENT HEWLETT PACKARD
Forensic anion solutions kit
Katalog # HEWL5064-8209
CAS nummer:  
Forensic anion solutions kit

Buffer, basic anion, for CE, pH 12,1, 50 ml, indirect UV-detection buffer, 1

CAS nummer:  
Price
888,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

This highly focused kit was developed specifically for the analysis of poisonous compounds, such as cyanide, azide, selenate, arsenate, and arsenite.

  • Requires minimal sample preparation

In cases of poisoning, analytical tools are needed to determine the identity of the toxins quickly and accurately. A rapid determination of anionic toxins in adulterated foods and beverages is possible using CE with indirect UV detection. Forensic and other anions can be detected within 15 minutes with minimal sample preparation.

Specifikationer

Specifikationer

  • Pakn:
    1
  • Beskrivelse:
    Buffer, basic anion, for CE, pH 12,1, 50 ml, indirect UV-detection buffer

Specifikationer