Forensic anion solutions kit
Forensic anion solutions kit for CE, 1
Katalog # HEWL5064-8208
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
This highly focused kit was developed specifically for the analysis of poisonous compounds, such as cyanide, azide, selenate, arsenate, and arsenite.
- Requires minimal sample preparation
In cases of poisoning, analytical tools are needed to determine the identity of the toxins quickly and accurately. A rapid determination of anionic toxins in adulterated foods and beverages is possible using CE with indirect UV detection. Forensic and other anions can be detected within 15 minutes with minimal sample preparation.
Specifikationer
Specifikationer
- Pakn:1
- Beskrivelse:Forensic anion solutions kit for CE