Capillary electrochromatography (CEC) capillaries
Product Details & Documents
Capillary electrochromatography is a hybrid of CE and LC and can be performed in the Agilent CE system. Using CE capillaries packed with LC stationary phases, CEC offers the loadability and selectivity of LC and the high efficiency of CE.
Using the high pressure capabilities of the Agilent CE system, both ends of the CEC capillary can be pressurised. This process prevents outgassing upon application of high voltage and significantly extends capillary lifetime.
Use CEC to improve resolution of solutes, which are difficult to resolve by HPLC, for hydrophobic solutes which cannot be solubilised in MEKC buffers, or for reduced sample and solvent consumption compared to HPLC.
The colour coding of the capillary (alignment stopper) and the interface makes it easy to combine the correct alignment interface with the capillary.
Specifikationer
- Beskrivelse:Standard packed CEC capillaries, C8, 3 μm, ID 100 μm
- Seq:3
- Samlet længde:335 mm
- Farvekode:grå
- Brugbar længde:250 mm
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



