Capillaries for LC systems
Capillaries, without fittings, PEEK, 48 mm, I.Ø 0,25 mm, to connect VWD to waste
Katalog # HEWL5062-8535
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifikationer
- Beskrivelse:Capillaries, without fittings, PEEK, 48 mm, I.Ø 0,25 mm, to connect VWD to waste
- Seq:68