Capillary column ferrules for gas chromatography
Specialty ferrules, 85% vespel, 15% graphite, with two holes, ferrule ID 0,5 mm, column ID 0,32 mm
Katalog # HEWL5062-3581
Leverandør: AGILENT HEWLETT PACKARD
Capillary column ferrules for gas chromatography
Katalog # HEWL5062-3581
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Documents
HEWL5062-3514_TDS_EN
Datasheet
Specifikationer
- Beskrivelse:Specialty ferrules, 85% vespel, 15% graphite, with two holes, ferrule ID 0,5 mm, column ID 0,32 mm
- Seq:17