Orders LinkContactUsLinkComponent
Capillary column ferrules for gas chromatography
Capillary column ferrules for gas chromatography
Specialty ferrules, 85% vespel, 15% graphite, witrh two holes, ferrule ID 0,4 mm, column ID 0,1, 0,2, 0,25 mm
Katalog # HEWL5062-3580
Leverandør:   AGILENT HEWLETT PACKARD
Capillary column ferrules for gas chromatography
Katalog # HEWL5062-3580
Capillary column ferrules for gas chromatography
Capillary column ferrules for gas chromatography
Specialty ferrules, 85% vespel, 15% graphite, witrh two holes, ferrule ID 0,4 mm, column ID 0,1, 0,2, 0,25 mm
Price
1.000,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
Documents
Specifikationer
  • Beskrivelse:
    Specialty ferrules, 85% vespel, 15% graphite, witrh two holes, ferrule ID 0,4 mm, column ID 0,1, 0,2, 0,25 mm
  • Seq:
    16