Orders LinkContactUsLinkComponent
Capillary column ferrules for gas chromatography
Capillary column ferrules for gas chromatography
85% vespel, 15% graphite ferrules, short, ferrule ID 0,5 mm, column ID 0,32 mm
Katalog # HEWL5062-3514
Leverandør:   AGILENT HEWLETT PACKARD
Capillary column ferrules for gas chromatography
Katalog # HEWL5062-3514
Capillary column ferrules for gas chromatography
Capillary column ferrules for gas chromatography
85% vespel, 15% graphite ferrules, short, ferrule ID 0,5 mm, column ID 0,32 mm
Price
642,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
Documents
Specifikationer
  • Beskrivelse:
    85% vespel, 15% graphite ferrules, short, ferrule ID 0,5 mm, column ID 0,32 mm
  • Seq:
    06