Capillary column ferrules for gas chromatography
High performance ferrules, 100% vespel, short, ferrule ID 0,5 mm, column ID 0,32 mm
Katalog # HEWL5062-3513
Leverandør: AGILENT HEWLETT PACKARD
Capillary column ferrules for gas chromatography
Katalog # HEWL5062-3513
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Documents
HEWL5062-3514_TDS_EN
Datablad
Specifikationer
- Beskrivelse:High performance ferrules, 100% vespel, short, ferrule ID 0,5 mm, column ID 0,32 mm
- Seq:14