Capillary column ferrules for gas chromatography
85% vespel, 15% graphite ferrules, short, ferrule ID 0,4 mm, column ID 0,45, 0,53 mm
HEWL5062-3512
Documents
HEWL5062-3514_TDS_EN
Datablad
- Beskrivelse:85% vespel, 15% graphite ferrules, short, ferrule ID 0,4 mm, column ID 0,45, 0,53 mm
- Seq:05