Orders LinkContactUsLinkComponent
Capillary column ferrules for gas chromatography
Capillary column ferrules for gas chromatography
High performance ferrules, 100% vespel, short, ferrule ID 0,8 mm, column ID 0,45, 0,53 mm
Katalog # HEWL5062-3511
Leverandør:   AGILENT HEWLETT PACKARD
Capillary column ferrules for gas chromatography
Katalog # HEWL5062-3511
Capillary column ferrules for gas chromatography
Capillary column ferrules for gas chromatography
High performance ferrules, 100% vespel, short, ferrule ID 0,8 mm, column ID 0,45, 0,53 mm
Price
kr642.00
Log in for availability
1 * 10 Items
Product Details & Documents
Documents
Specifications
  • Beskrivelse:
    High performance ferrules, 100% vespel, short, ferrule ID 0,8 mm, column ID 0,45, 0,53 mm
  • Seq:
    13