Capillary column ferrules for gas chromatography
High performance ferrules, 100% vespel, short, ferrule ID 0,8 mm, column ID 0,45, 0,53 mm
Katalog # HEWL5062-3511
Leverandør: AGILENT HEWLETT PACKARD
Capillary column ferrules for gas chromatography
Katalog # HEWL5062-3511
Leverandør: AGILENT HEWLETT PACKARD
Jump to:
Product Details & Documents
Documents
HEWL5062-3514_TDS_EN
Datasheet
Specifications
- Beskrivelse:High performance ferrules, 100% vespel, short, ferrule ID 0,8 mm, column ID 0,45, 0,53 mm
- Seq:13