Capillary column ferrules for gas chromatography
Preconditioned 85% vespel, 15% graphite ferrules, long, ferrule ID 0,4 mm, column ID 0,1, 0,2, 0,25 mm
Katalog # HEWL5062-3508
Leverandør: AGILENT HEWLETT PACKARD
Capillary column ferrules for gas chromatography
Katalog # HEWL5062-3508
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Preconditioned 85% vespel, 15% graphite ferrules, long, ferrule ID 0,4 mm, column ID 0,1, 0,2, 0,25 mm
- Seq:11