Capillary column ferrules for gas chromatography
Preconditioned 85% vespel, 15% graphite ferrules, long, ferrule ID 0,3 mm, column ID 0,1 mm
Katalog # HEWL5062-3507
Leverandør: AGILENT HEWLETT PACKARD
Capillary column ferrules for gas chromatography
Katalog # HEWL5062-3507
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Documents
HEWL5062-3514_TDS_EN
Datablad
Specifikationer
- Beskrivelse:Preconditioned 85% vespel, 15% graphite ferrules, long, ferrule ID 0,3 mm, column ID 0,1 mm
- Seq:10