Orders LinkContactUsLinkComponent
Capillary column ferrules for gas chromatography
Capillary column ferrules for gas chromatography
Preconditioned 85% vespel, 15% graphite ferrules, long, ferrule ID 0,5 mm, column ID 0,32 mm
Catalog # HEWL5062-3506
Capillary column ferrules for gas chromatography
Catalog # HEWL5062-3506
Capillary column ferrules for gas chromatography
Capillary column ferrules for gas chromatography
Preconditioned 85% vespel, 15% graphite ferrules, long, ferrule ID 0,5 mm, column ID 0,32 mm
Price
702,00 kr
Log in for availability
1 * 10 Items
Product Details & Documents
Documents
Specifications
  • Beskrivelse:
    Preconditioned 85% vespel, 15% graphite ferrules, long, ferrule ID 0,5 mm, column ID 0,32 mm
  • Seq:
    09