Capillary column ferrules for gas chromatography
Preconditioned 85% vespel, 15% graphite ferrules, long, ferrule ID 0,5 mm, column ID 0,32 mm
Catalog # HEWL5062-3506
Supplier: AGILENT HEWLETT PACKARD
Capillary column ferrules for gas chromatography
Catalog # HEWL5062-3506
Supplier: AGILENT HEWLETT PACKARD
Jump to:
Product Details & Documents
Documents
HEWL5062-3514_TDS_EN
Datasheet
Specifications
- Beskrivelse:Preconditioned 85% vespel, 15% graphite ferrules, long, ferrule ID 0,5 mm, column ID 0,32 mm
- Seq:09