Capillaries for LC systems
Restriction capillaries, stainless steel, with 1/16” female connector on both ends, 2000 mm, I.Ø 0,12 mm
Catalog # HEWL5022-2159
Supplier: AGILENT HEWLETT PACKARD
Jump to:
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifications
- Beskrivelse:Restriction capillaries, stainless steel, with 1/16” female connector on both ends, 2000 mm, I.Ø 0,12 mm
- Seq:108