Orders LinkContactUsLinkComponent
Capillaries for LC systems
Capillaries for LC systems
Restriction capillaries, stainless steel, with 1/16” female connector on both ends, 2000 mm, I.Ø 0,12 mm
Catalog # HEWL5022-2159
Capillaries for LC systems
Catalog # HEWL5022-2159
Capillaries for LC systems
Capillaries for LC systems
Restriction capillaries, stainless steel, with 1/16” female connector on both ends, 2000 mm, I.Ø 0,12 mm
Price
1.210,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifications
  • Beskrivelse:
    Restriction capillaries, stainless steel, with 1/16” female connector on both ends, 2000 mm, I.Ø 0,12 mm
  • Seq:
    108