Capillary column ferrules for gas chromatography
Graphpak capillary column connectors, column ID 0,53 mm, connector ID 0,7 mm
Katalog # HEWL5021-7134
Leverandør: AGILENT HEWLETT PACKARD
Capillary column ferrules for gas chromatography
Katalog # HEWL5021-7134
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Documents
HEWL5062-3514_TDS_EN
Datablad
Specifikationer
- Beskrivelse:Graphpak capillary column connectors, column ID 0,53 mm, connector ID 0,7 mm
- Seq:27