Orders LinkContactUsLinkComponent
Capillaries for LC systems
Capillaries for LC systems
Capillaries, without fittings, red, stainless steel, 400 mm, I.Ø 0,12 mm, to connect column to VWD
Katalog # HEWL5021-1823
Leverandør:   AGILENT HEWLETT PACKARD
Capillaries for LC systems
Katalog # HEWL5021-1823
Capillaries for LC systems
Capillaries for LC systems
Capillaries, without fittings, red, stainless steel, 400 mm, I.Ø 0,12 mm, to connect column to VWD
Price
351,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifikationer
  • Beskrivelse:
    Capillaries, without fittings, red, stainless steel, 400 mm, I.Ø 0,12 mm, to connect column to VWD
  • Seq:
    72