Capillaries for LC systems
Capillaries, without fittings, red, stainless steel, 105 mm, I.Ø 0,12 mm, to connect column to VWD
Katalog # HEWL5021-1820
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifikationer
- Beskrivelse:Capillaries, without fittings, red, stainless steel, 105 mm, I.Ø 0,12 mm, to connect column to VWD
- Seq:69