Orders LinkContactUsLinkComponent
Capillary column ferrules for gas chromatography
Capillary column ferrules for gas chromatography
Finger tight column nut, small, for 320 μm columns and smaller
Katalog # HEWL5020-8292
Leverandør:   AGILENT HEWLETT PACKARD
Capillary column ferrules for gas chromatography
Katalog # HEWL5020-8292
Capillary column ferrules for gas chromatography
Capillary column ferrules for gas chromatography
Finger tight column nut, small, for 320 μm columns and smaller
Price
247,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Documents
Specifikationer
  • Beskrivelse:
    Finger tight column nut, small, for 320 μm columns and smaller
  • Seq:
    57