Mass flow controllers and restrictors
Mass flow controller, with 0 - 400 ml/min restrictor, for use with capillary column inlets
Catalog # HEWL19362-60575
Supplier: AGILENT HEWLETT PACKARD
Jump to:
Product Details & Documents
Specifications
- Beskrivelse:Mass flow controller, with 0 - 400 ml/min restrictor, for use with capillary column inlets
- Seq:02