Split/splitless inlet seals
Septum retainer nut
Catalog # HEWL18740-60835
Supplier: AGILENT HEWLETT PACKARD
Jump to:
Product Details & Documents
These inlet seals have been developed to ensure that you have a consistent and inert surface to properly seal the inlet and prevent sample degradation.
- Very reproducible smooth surface, eliminating traces of machining grooves that can be the source of minute leaks
- High quality, leak-free seal
Delivery information: An inlet washer is provided with each inlet seal.
Specifications
- Beskrivelse:Septum retainer nut
- Seq:9