Preventive maintenance kit for LC/MS
Oil mist filter element, for E1M18/E2M28, LC/MS
Catalog # HEWL1535-4970
Supplier: AGILENT HEWLETT PACKARD
Jump to:
Product Details & Documents
Specifications
- Til:LC/MS
- Beskrivelse:Oil mist filter element, for E1M18/E2M28
- Seq:22