Orders LinkContactUsLinkComponent
Capillaries for LC systems
Capillaries, pre-swaged, blue, stainless steel, 130 mm, I.Ø 0,25 mm, to connect pump to autosampler
Katalog # HEWL01090-87308
Leverandør:   AGILENT HEWLETT PACKARD
Capillaries for LC systems
Katalog # HEWL01090-87308
Capillaries for LC systems
Capillaries for LC systems
Capillaries, pre-swaged, blue, stainless steel, 130 mm, I.Ø 0,25 mm, to connect pump to autosampler
Price
1.360,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifikationer
  • Beskrivelse:
    Capillaries, pre-swaged, blue, stainless steel, 130 mm, I.Ø 0,25 mm, to connect pump to autosampler
  • Seq:
    46