Capillaries for LC systems
Capillaries, 1 end pre-swaged, blue, stainless steel, 700 mm, I.Ø 0,25 mm, to connect pump to universal
Katalog # HEWL01018-67305
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifikationer
- Beskrivelse:Capillaries, 1 end pre-swaged, blue, stainless steel, 700 mm, I.Ø 0,25 mm, to connect pump to universal
- Seq:03