Orders LinkContactUsLinkComponent
CrossLab Bottle Head Assemblies for HPLC
CrossLab Bottle Head Assemblies for HPLC
Solvent inlet filter, stainless steel
Katalog # HEWL01018-60025
Leverandør:   AGILENT HEWLETT PACKARD
CrossLab Bottle Head Assemblies for HPLC
Katalog # HEWL01018-60025
CrossLab Bottle Head Assemblies for HPLC
CrossLab Bottle Head Assemblies for HPLC
Solvent inlet filter, stainless steel
Price
1.090,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
All the parts required to connect solvent bottles to your HPLC instrument.
  • Instrument specific ready-to-use bottle head assemblies available
  • Special ultraclean tubing for MS applications available
Agilent bottle head assemblies contain all the parts that are required to connect solvent bottles to your HPLC instrument. Each assembly contains a solvent inlet filter, tubing, a bottle cap, and fittings.
Specifikationer
  • Beskrivelse:
    Solvent inlet filter, stainless steel
  • Seq:
    24