CrossLab Bottle Head Assemblies for HPLC
Solvent inlet filter, stainless steel
Katalog # HEWL01018-60025
Leverandør: AGILENT HEWLETT PACKARD
CrossLab Bottle Head Assemblies for HPLC
Katalog # HEWL01018-60025
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
All the parts required to connect solvent bottles to your HPLC instrument.
- Instrument specific ready-to-use bottle head assemblies available
- Special ultraclean tubing for MS applications available
Agilent bottle head assemblies contain all the parts that are required to connect solvent bottles to your HPLC instrument. Each assembly contains a solvent inlet filter, tubing, a bottle cap, and fittings.
Specifikationer
- Beskrivelse:Solvent inlet filter, stainless steel
- Seq:24