Orders LinkContactUsLinkComponent
Capillary column ferrules for gas chromatography
Graphite ferrule, 1/16"
Katalog # HEWL0100-1326
Leverandør:   AGILENT HEWLETT PACKARD
Capillary column ferrules for gas chromatography
Katalog # HEWL0100-1326
Capillary column ferrules for gas chromatography
Capillary column ferrules for gas chromatography
Graphite ferrule, 1/16"
Price
381,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Graphite ferrule, 1/16"
  • Seq:
    45