Jars with base, knobbed lid
100 mm, 100 mm
Katalog # 216-4293
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
Made of glass, with glass or stainless steel lid.
- Good chemical resistance
- Sturdy
- Easy to clean
Suitable for storing dressing material, cotton, clinical thermometers, forceps and other instruments.
Specifikationer
- Udv.Ø:100 mm
- Højde:100 mm