Orders LinkContactUsLinkComponent
Electrolysis apparatus acc.Hoffmann
Electrolysis apparatus acc.Hoffmann
Water decomposition device
Katalog # 761-0093
Electrolysis apparatus acc.Hoffmann
Katalog # 761-0093
Electrolysis apparatus acc.Hoffmann
Electrolysis apparatus acc.Hoffmann
Water decomposition device
Price
3.610,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
According to Hoffmann, with two city dishes, 0 to 30 ml of grading graded 1/10 ml.
 : Supplied with metal tripod and two clamps.
Specifikationer
  • Beskrivelse:
    Water decomposition device
  • Seq:
    01
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Water decomposer, only the glass details
    Water decomposer, only the glass details
    761-0094
    Log ind for tilgængelighed
    1 * 1 Items - 1.530,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Platinum electrodes
    Platinum electrodes
    761-0095
    Log ind for tilgængelighed
    1 * 1 Items - 501,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Kul-elektrode
    Kul-elektrode
    761-0096
    Log ind for tilgængelighed
    1 * 1 Items - 172,00 kr