Instrument dishes
200x100x50 mm
Katalog # 216-1403
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
Thick-walled glass.
- Can be sterilised in hot air up to 180 °C
- Robust
- Stainless steel lids are available separately
Multipurpose dishes with vertical walls for a variety of applications in the laboratory, field use and industry.
Specifikationer
- Seq:03
- B×D×H:200x100x50 mm