Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Graduated pipettes, with piston

25 ml, 0,1 ml, ±0,150 ml, hvid
  HECH42548056
Graduated pipettes, with piston
  HECH42548056
Graduated pipettes, with piston
Graduated pipettes, with piston

25 ml, 0,1 ml, ±0,150 ml, hvid

 
435,00 kr
   
1 * 1 Items
 

Safety measuring pipettes with interchangeable ground-in piston, standard type.

  • Tolerance 1,5 times higher than the official tolerance of class AS
  • Fused amber graduation 'ASSISTENT-PERMANENT'
  • Zero at top, calibrated
  • Fire-polished tip

 : Each pipette is packed in a plastic tube.

 
  • Farvekode:
    hvid
  • Kapacitet:
    25 ml
  • Seq:
    05
  • Inddeling:
    0,1 ml
  • Tolerance:
    ±0,150 ml