Top tilt measures
25 ml
Katalog # HECH41877012
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
Automatic tilt measures, made of DURAN® borosilicate glass 3.3, with standard ground cone NS 29/32. Supplied without reservoir bottle.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Specifikationer
- Kapacitet:25 ml
- Seq:04