Orders LinkContactUsLinkComponent
Top tilt measures
Top tilt measures
25 ml
Katalog # HECH41877012
Top tilt measures
Katalog # HECH41877012
Top tilt measures
Top tilt measures
25 ml
Price
356,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Automatic tilt measures, made of DURAN® borosilicate glass 3.3, with standard ground cone NS 29/32. Supplied without reservoir bottle.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Specifikationer
  • Kapacitet:
    25 ml
  • Seq:
    04