Orders LinkContactUsLinkComponent
Top tilt measures
Top tilt measures
100 ml
Catalog # HECH41877018
Top tilt measures
Catalog # HECH41877018
Top tilt measures
Top tilt measures
100 ml
Price
801,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Automatic tilt measures, made of DURAN® borosilicate glass 3.3, with standard ground cone NS 29/32. Supplied without reservoir bottle.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Specifications
  • Kapacitet:
    100 ml
  • Seq:
    06