Staining racks
Staining rack, extendable, Plexiglas®
Katalog # 631-0336
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
Staining rack, Plexiglas®, in the form of an extendable bridge, that can be fixed in the horizontal or at an inclined position. Suitable for trays or dishes of 380 to 620 mm in length.
Specifikationer
- Beskrivelse:Staining rack, extendable, Plexiglas®