Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Mixing spatulas, double ended

Mixing spatulas, double ended

180 mm, 6 mm
Katalog # 100-0512
Leverandør:   HAMMACHER KARL
Mixing spatulas, double ended
Katalog # 100-0512
Leverandør:  HAMMACHER KARL
Mixing spatulas, double ended
Mixing spatulas, double ended

180 mm, 6 mm

Price
553,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Double ended mixing spatulas made from WIRONIT steel.

Specifikationer
  • Længde:
    180 mm
  • Seq:
    09
  • Bladbredde:
    6 mm