Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Mixing spatulas, double ended

Mixing spatulas, double ended

180 mm, 5 mm
Catalog # 231-2361
Supplier:   HAMMACHER KARL
Mixing spatulas, double ended
Catalog # 231-2361
Supplier:  HAMMACHER KARL
Mixing spatulas, double ended
Mixing spatulas, double ended

180 mm, 5 mm

Price
553,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Double ended mixing spatulas made from WIRONIT steel.

Specifications
  • Længde:
    180 mm
  • Seq:
    08
  • Bladbredde:
    5 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.