Orders LinkContactUsLinkComponent
Cover glass forceps
Cover glass forceps
105 mm
Catalog # 232-2200
Supplier:   HAMMACHER KARL
Cover glass forceps
Catalog # 232-2200
Supplier:  HAMMACHER KARL
Cover glass forceps
Cover glass forceps
105 mm
Price
489,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Wironit steel (18/12 stainless steel with 18% chrome and 10 - 12% nickel).
  • For microscopic slides
  • Resistant to corrosion
  • Anti-magnetic
Specifications
  • Længde:
    105 mm