Forceps, teethed
120 mm
Katalog # 232-0067
Leverandør: HAMMACHER KARL
Gå til:
Product Details & Documents
WIRONIT steel (18/12 stainless steel with 18% chrome and 10 - 12 % nickel)
- Teethed, straight
- Resistant to corrosion
- Anti-magnetic
Specifikationer
- Længde:120 mm