Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Double spatulas

Double-ended, both ends spoon-formed (spoon size 10 mm), 210 mm
  HAMMHSN122-21
Double spatulas
  HAMMHSN122-21
Double spatulas
Double spatulas

Double-ended, both ends spoon-formed (spoon size 10 mm), 210 mm

 
104,00 kr
   
1 * 1 Items
 

Double-ended spatulas made from stainless steel.

 
  • Beskrivelse:
    Double-ended, both ends spoon-formed (spoon size 10 mm)
  • Længde:
    210 mm
  • Seq:
    03