Orders LinkContactUsLinkComponent
Disposable tip adapters
Disposable tip adapters
Disposable tip adapter, CTFE, 1 ml
Catalog # HAMI31331
Supplier:   HAMILTON BONADUZ
Disposable tip adapters
Catalog # HAMI31331
Supplier:  HAMILTON BONADUZ
Disposable tip adapters
Disposable tip adapters
Disposable tip adapter, CTFE, 1 ml
Price
565,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
These adapters enable the connection of a standard 1 ml or 200 µl disposable pipette tip to the end of a Hamilton syringe. The adapter connects to standard Luer Tip (LT) and PTFE Luer Lock (TLL) syringes. When used in conjunction the PB600-1 and the disposable tip adapters can be used as a small volume repeating pipette.
  • Ideal for eliminating the possibility for cross contamination
Specifications
  • Beskrivelse:
    Disposable tip adapter, CTFE, 1 ml
  • Seq:
    02